missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCl 0.01D Conductivity Standards, Hach Lange™
Description
- Features a concentration of 1408 μS/cm ± 0.5 % at 25°C
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- According to OIML, traceable to NIST
- Standard solution verified by Reference Materials Laboratory
- Shelf life of 24 months from production date
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Specifications
| Methods | Standard solution verified by Reference Materials Laboratory |
| Analyte or Component Names | KCL |
| Conductivity | 1408μS/cm ± 0.5% at 25°C |
| Physical Form | Liquid |
| Concentration | 1408 μS/cm ±0.5% at 25°C |
| Element | KCl |
| Quantity | 500 mL |