missing translation for 'onlineSavingsMsg'
Learn More

Kauri-Butanol Solution, Spectrum™ Chemical

Catalog No. 18611154
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18611154 500 mL
18611153 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611154 Supplier Spectrum Chemical Mfg Cor Supplier No. K101500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Kauri-Butanol Solution
CAS 77465-88-4, 71-36-3, 9000-14-0
CAS Max % 84.6, 15.4
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.