missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kauri-Butanol Solution, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Kauri-Butanol Solution |
| CAS | 77465-88-4, 71-36-3, 9000-14-0 |
| CAS Max % | 84.6, 15.4 |
| Grade | Chemical Solution |
| Packaging | Amber Glass Bottle |
| Quantity | 100 mL |
| Physical Form | Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.