missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS
Description
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is a 10μg/ml N-desacetyl-N-methylocolchicine (Colcemid™) solution made in Phosphate Buffered Saline (PBS). It is used for arresting cells in metaphase so the chromosomes can be separated for cytogenetic studies and in vitro diagnostic procedures. Colcemid™ prevents spindle formation during mitosis, causing metaphase arrest. The mechanism of action is similar to that of colchicine, but with lower mammalian toxicity. The complete formulation is available.
Product Use
For in vitro diagnostic use.
cGMP Manufacturing and Quality System
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Product Use
For in vitro diagnostic use.
cGMP Manufacturing and Quality System
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Specifications
Specifications
| Content And Storage | Storage conditions: 2 – 8°C Shipping conditions: Ambient Shelf life: 18 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Form | Liquid |
| Product Type | Chromosome Analysis Media |
| Sterility | Sterile-filtered |
| For Use With (Application) | Clinical Research, Translational Research |
| pH | 6 to 8 |
| Product Line | KaryoMAX |
| Quantity | 10 mL |
| Shipping Condition | Room Temperature |