missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Reagent Solution A, for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol), Honeywell™ Fluka™

Catalog No. p-7122085
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000881 1 L
60046639 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000881 Supplier Honeywell Chemicals Supplier No. 361161L
Only null left

for pyridine-based volumetric Karl Fischer titration (pyridine, sulfur dioxide, methanol)

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations

Specifications

Chemical Name or Material Karl Fischer reagent solution A
Density 1.01g/mL
Quantity 1 L
Flash Point 16°C (60.8°F)
Molecular Formula CH4O
Packaging Glass bottle
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Show More Show Less