missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. K2000212
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K2000212 2.5 L
K20001 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K2000212 Supplier Fisher Chemical Supplier No. K200017

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free

Specifications

Chemical Name or Material Aqualine™ Complete 5
Name Note Karl Fischer Reagent
Color Brown
CAS 7446-09-5, 288-32-4, 111-90-0, 7553-09-5
Physical Form Liquid
Quantity 2.5 L
Packaging Amber glass bottle
MDL Number 81734