Learn More
SW Sustainability Solutions KarbonHex KX80 Purpose Built Cut-Resistant Gloves with Oeko-Tex Safety Certification
Designed to not accumulate static electricity, purpose built gloves feature cut resistance and touchscreen compatibility. Nitrile dipped palm coating offers performance grip, extreme dexterity, and precise handling of small components in wet, oily and slippery conditions.
Supplier:  SW Sustainability Solutions C442804
 View more versions of this product
Catalog No. 17441174
Only null left
Explore available promotions
Description
Description
  • Oeko-Tex certified for customer confidence and high product safety.
  • Super lightweight close fitting 18 G composite knit shell provides exceptional fit, feel and all-round cut protection.
  • Sandy foam nitrile palm coating provides precise oil and wet grip handling.
  • Fully touchscreen compatible.
  • Elastic knit wrist ensures a comfortable and secure fit.
Specifications
Specifications
ANSI-ISEA 105-16
Seamless Knit
Nitrile Foam
ANSI A4
Yes
Cut-resistant Gloves
EN388:4X42D
Gray
Sandpatch
Seamless Lining
ANSI/ISEA 105-16 Level 3
Straight
10.2 in.
EN388:2016
Elastic
Black, Gray
Specific
HPPE
Large
Unisex
Knit
Composite Fibers
C442804
C44280
18 G
25.9 cm
Product Suggestions
Product Suggestions