missing translation for 'onlineSavingsMsg'
Learn More

kappa-Carrageenan, TCI America™

Catalog No. C180425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C180425G 25 g
C1804500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C180425G Supplier TCI America Supplier No. C180425G

Chemical Identifiers

CAS 11114-20-8
Molecular Formula C24H36O25S2-2
Molecular Weight (g/mol) 788.647
MDL Number MFCD00151514
InChI Key ZNOZWUKQPJXOIG-XSBHQQIPSA-L
Synonym kappa-carrageenan, kappa-carrageenans, 1->4-3,6-anhydro-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
PubChem CID 11966249
IUPAC Name [(2R,3S,4R,5R,6S)-6-[[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-[[(1R,3R,4R,5R,8S)-8-[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-sulfonatooxyoxan-2-yl]oxy-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-3-yl]oxy]-5-hydroxy-2-(
SMILES C1C2C(C(O1)C(C(O2)O)O)OC3C(C(C(C(O3)CO)OS(=O)(=O)[O-])OC4C(C5C(C(O4)CO5)OC6C(C(C(C(O6)CO)OS(=O)(=O)[O-])O)O)O)O

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material kappa-Carrageenan
RTECSNumber FI0703000
TSCA No