missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Powder, USP, Spectrum™ Chemical

Catalog No. p-7157650
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603622 500 g Poly Bottle
18603624 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603622 Supplier Spectrum Chemical Mfg Cor Supplier No. KA105500GM
Only null left

KA105, 1332-58-7

Spectrum™ Chemical Kaolin, Powder, USP is used to treat diarrhea and to relieve soreness and swelling inside the mouth caused by radiation treatments. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

CAS Max % 100%
Quantity 500 g
Grade USP
Loss on Ignition 15%
Packaging Poly Bottle