accessibility menu, dialog, popup

UserName

H2OH2OOAlOSiOOSiOOAlO

CAS RN 1332-58-7

IUPAC Name: dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
Synonyms: halloysite kaolin 白陶土
Molecular Weight (g/mol): 258.16
Molecular Formula: Al2H4O9Si2
InChi Key: NLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILES: O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
Grade 
  • (6)
  • (3)
  • (2)
Physical Form 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (6)
  • (3)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)