missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaolin, Powder, FCC, Spectrum™ Chemical
K1004, 1332-58-7
Supplier: Spectrum Chemical Mfg Cor K100425KGCS4
Description
Spectrum™ Chemical Kaolin, Powder, FCC is used as a whitener in toothpaste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 1332-58-7 | |
| 4 x 2.5 kg | |
| MFCD00062311 | |
| O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O | |
| 258.16 | |
| Poly Pail |
| 100% | |
| Al2H4O9Si2 | |
| NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
| dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate | |
| 15% |
Chemical Identifiers
| 1332-58-7 | |
| 258.16 | |
| NLYAJNPCOHFWQQ-UHFFFAOYSA-N | |
| O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O |
| Al2H4O9Si2 | |
| MFCD00062311 | |
| dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate |