missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK2/SIP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine
Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK2 (KN motif and ankyrin repeat domain-containing protein 2), was initially identified as a steroid receptor coactivator (SRC) interacting protein (SIP) that could sequester steroid receptor coactivators in the cytoplasm. More recent experiments have shown that while KANK2 is widely expressed, in kidney podocytes, KANK2 localizes to foot processes, suggesting that KANK2 may contribute to controlling actin dynamics in podocyte foot processes.
Specifications
Specifications
| Antigen | KANK2/SIP |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | KANK2 |
| Gene Accession No. | Q63ZY3 |
| Gene Alias | KANK2, KN motif and ankyrin repeat domains 2, SRC-interacting protein, SIP, matrix-remodeling-associated protein 3, MXRA3, Ankyrin repeat domain-containing protein 25, ANKRD25 |
| Gene Symbols | KANK2 |
| Show More |