missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Sulfate, Ultrapure

CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

$103.15 - $435.30

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin A
PubChem CID 134129479
IUPAC Name (2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AAJ1792406
View Documents Promotion Details
N/A 5 g Plastic Bottle
Each for $103.15
Only null left
 
AAJ1792414
View Documents Promotion Details
N/A 25 g Plastic Bottle
Each for $435.30
Only null left
 
Description

Description

A selectable marker for cosmid vectors used in transfection of mammalian cells.

Mode of Action: Binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selectable marker for cosmid vectors used in transfection of mammalian cells
  • Mode of Action: Binds to ribosomal components and inhibits protein synthesis
  • Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in periplasmic space
Specifications

Chemical Identifiers

25389-94-0
582.575
OOYGSFOGFJDDHP-IZQIRFRQSA-N
134129479
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
C18H38N4O15S
MFCD00070253
Kanamycin A
(2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid

Specifications

Kanamycin Sulfate
25389-94-0
MFCD00070253
OOYGSFOGFJDDHP-IZQIRFRQSA-N
(2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
134129479
Kanamycin sulfate
Powder
C18H38N4O15S
Kanamycin A
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
582.575
582.58
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H360FD

EINECSNumber : 246-933-9

RTECSNumber : NZ3225030

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only