missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin monosulfate, MP Biomedicals™

Catalog No. ICN15002905
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15002905 5 g
ICN15002925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15002905 Supplier MP Biomedicals Inc Supplier No. 0215002905

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin sulfate salt, Kanamycin A
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Monosulfate
Physical Form Powder
Color White
Quantity 5 g
Formula Weight 582.6
Chemical Name or Material Kanamycin monosulfate