missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin monosulfate, Cell Culture Grade

Catalog No. AAJ6066803
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6066803 1 g
AAJ6066806 5 g
AAJ6066809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6066803 Supplier Thermo Scientific Chemicals Supplier No. J6066803

Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

Kanamycin monosulfate, is used as an aminoglycoside antibiotic. Kanamycin can be used as a selection agent in several types of isolation media.Kanamycin sulfate from Streptomyces kanamyceticus has been used in tissue culture media. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Kanamycin monosulfate is the sulfate salt form of kanamycin A

• It was isolated from Streptomyces kanamyceticus

• This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

• Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species

• This compound is also involved in translating prokaryotic ribosomes

• In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin A
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Monosulfate
Physical Form Powder
Color White
Quantity 1 g
Merck Index 14,5281
Solubility Information Soluble in water at 10mg/ml
Formula Weight 582.58
Grade Cell Culture
Chemical Name or Material Kanamycin monosulfate
Hazard Category H317-H334-H360FD
Hazard Statement GHS H Statement
H334-H360-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 246-933-9
RTECSNumber NZ3225030
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only