missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Kallikrein 14 ELISA Kit

Catalog No. EHKLK14
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHKLK14 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHKLK14 Supplier Invitrogen™ Supplier No. EHKLK14
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The Human Kallikrein 14 (Hu KLK14) ELISA quantitates Hu KLK14 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu KLK14. Principle of the method The Human KLK14 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.

Specifications

Accession Number Q9P0G3
Assay Range 1.5-500 pg/mL
Assay Sensitivity 1.5 pg/mL
Calibration Range 1.5 pg/mL to 500 pg/mL
Content And Storage Pre-coated 96-well plate, standard, detector antibody, streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. May be stored at 2–8°C for up to 6 months from the date of shipment.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family GPCR (G-Protein Coupled Receptors)
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human KLK14. Other species not determined.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14; KLK-L6
Gene ID (Entrez) 43847
Gene Symbol Klk14
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases hK14, Kallikrein-like protein 6, KLK-L6
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Biomarker-Related Pathways
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym KLK-L6
Target Kallikrein 14
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less