missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kallikrein 13 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA116656

Catalog No. APPA6656


Only null left

Description

Description

Kallikrein 13 Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ELISA

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L4, is expressed in a variety of tissues including prostate, salivary gland, breast, and testis. Its expression is regulated by steroid hormones in breast cancer cell lines may be involved in the pathogenesis and/or progression of breast cancer.
Specifications

Chemical Identifiers

Kallikrein 13
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
KLK13
DKFZp586J1923; Egfbp-2; kallikrein 13; kallikrein related peptidase 13; kallikrein related-peptidase 13; kallikrein-13; kallikrein-like gene 4; kallikrein-like protein 4; kallikrein-related peptidase 13; KLK13; KLK-13; KLKL4; KLK-L4; mGk-13
KLK13
Primary
Antigen affinity chromatography
1 mg/mL
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.02% sodium azide
Q9UKR3
26085
Residues 262-277 of the human KLK-L4 protein.
Antibody

Specifications

Kallikrein 13
Polyclonal
Unconjugated
KLK13
DKFZp586J1923; Egfbp-2; kallikrein 13; kallikrein related peptidase 13; kallikrein related-peptidase 13; kallikrein-13; kallikrein-like gene 4; kallikrein-like protein 4; kallikrein-related peptidase 13; KLK13; KLK-13; KLKL4; KLK-L4; mGk-13
Rabbit
Antigen affinity chromatography
RUO
26085
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UKR3
KLK13
Residues 262-277 of the human KLK-L4 protein.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions