missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol Hydrate 97.0+%, TCI America™

Catalog No. p-7131744
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K0018100MG 100 mg
K00181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K0018100MG Supplier TCI America Supplier No. K0018100MG
Only null left

Chemical Identifiers

CAS 520-18-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.24
MDL Number MFCD00016938
InChI Key IYRMWMYZSQPJKC-UHFFFAOYSA-N
Synonym kaempferol, kaempherol, kempferol, populnetin, rhamnolutein, robigenin, trifolitin, pelargidenolon, rhamnolutin, swartziol
PubChem CID 5280863
ChEBI CHEBI:28499
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 286.24
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Kaempferol Hydrate
RTECSNumber LK9275200
TSCA No