Learn More
JWH-073 N-(3-hydroxybutyl) Metabolite Solution, 100 μg/mL in Methanol, Ampule of 1 mL, Certified Reference Material, Cerilliant™
Supplier: Sartorius Corporation LAB1X2N1
Description
- The major and most prominent urinary metabolite of JWH-073, a synthetic cannabinoid found in several versions of the herbal mixture Spice.
- Our certified Spice cannabinoid solution standards are DEA-exempt allowing laboratories to order these standards without DEA paperwork.
Specifications
| 100 μg/mL | |
| JWH-073 N-(3-hydroxybutyl) Metabolite | |
| Ampule | |
| Liquid Chromatography (LC); Gas Chromatography (GC) |