Learn More
JWH-018 4-Hydroxypentyl Metabolite Solution, 100 μg/mL in Methanol, Ampule of 1 mL, Certified Reference Material, Cerilliant™
Supplier: Sartorius Corporation PMP01PXKR013
Description
- Also known as the Omega minus one-OH metabolite of JWH-018, the 4-Hydroxypentyl analog is the major urinary metabolite of JWH-018, a synthetic cannabinoid found in several versions of the herbal mixture Spice.
Specifications
| 100 μg/mL | |
| JWH-018 N-(4-hydroxypentyl) Metabolite | |
| Ampule | |
| Liquid Chromatography (LC); Gas Chromatography (GC) |