Learn More
JWH-018 4-Hydroxypentyl Metabolite-D5 (indole-D5) Solution, 100 μg/mL in Methanol, Ampule of 1 mL, Cerilliant™
Supplier: Sartorius Corporation EVO1X2N1C
Description
- A stable-labeled internal standard of JWH-018 4-Hydroxypentyl metabolite, also known as the Omega minus one-OH metabolite of JWH-018.
- The 4-Hydroxypentyl analog is the major urinary metabolite of JWH-018, a synthetic cannabinoid found in the herbal mixture Spice.
Specifications
| 100 μg/mL | |
| JWH-018 N-(4-hydroxypentyl) Metabolite-D5 (indole-D5) | |
| Ampule | |
| Liquid Chromatography (LC); Gas Chromatography (GC) |