missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Safely store hazardous materials with the Justrite Sure-Grip™ EX Hazardous Material Steel Safety Cabinet.

Supplier:  Justrite™ 863028

 View more versions of this product

Catalog No. 19159700


Only null left

Description

Description

  • Features double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet
  • Automatic self-close doors shuts and latches when fusible link melts at 74°C under fire conditions
  • Separate polyethylene liner for bottom sump to resist chemicals
  • Obstruction-free access to contents by concealed self-closing mechanism
  • Includes extra polyethylene work tray secured to cabinet top for handy work surface
  • Chemical resistance offered by epoxy baked-on powder-coat finish on inside and out surface
  • Comes with Hazardous - Keep Fire Away label, and pack of 10 application-specific labels to apply to cabinet to indicate its contents
  • Available in various capacities
  • Color: Royal blue

Compliance

NFPA 1, NFPA 400 Hazardous Material Code, and the International Fire Code

Warranty

Ten-year

Specifications

Chemical Identifiers

Royal Blue
Combi Safety Storage Cabinet
2

Specifications

14.6 x 39.5 x 39.6 in.
Combi Safety Storage Cabinet
2
1
18 x 43 x 44 in.
30 gal. (114 L)
Royal Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions