missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Catalog No. p-4831683
Change view
Click to view available options
Load Bearing Capacity:
30 gal. (114 L)
45 gal. (170 L)
No. of Shelves:
1
2
Dimensions (D x W x H):
18 x 43 x 44 in.
18 x 43 x 65 in.
Dimensions (D x W x H) Interior:
14.6 x 39.5 x 39.6 in.
14.6 x 39.5 x 60.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity No. of Shelves Dimensions (D x W x H) Dimensions (D x W x H) Interior
19159701 45 gal. (170 L) 2 18 x 43 x 65 in. 14.6 x 39.5 x 60.5 in.
19159700 30 gal. (114 L) 1 18 x 43 x 44 in. 14.6 x 39.5 x 39.6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19159701 Supplier Justrite™ Supplier No. 864528
Only null left

Safely store hazardous materials with the Justrite Sure-Grip™ EX Hazardous Material Steel Safety Cabinet.

  • Features double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet
  • Automatic self-close doors shuts and latches when fusible link melts at 74°C under fire conditions
  • Separate polyethylene liner for bottom sump to resist chemicals
  • Obstruction-free access to contents by concealed self-closing mechanism
  • Includes extra polyethylene work tray secured to cabinet top for handy work surface
  • Chemical resistance offered by epoxy baked-on powder-coat finish on inside and out surface
  • Comes with Hazardous - Keep Fire Away label, and pack of 10 application-specific labels to apply to cabinet to indicate its contents
  • Available in various capacities
  • Color: Royal blue

Compliance

NFPA 1, NFPA 400 Hazardous Material Code, and the International Fire Code

Warranty

Ten-year

Specifications

Dimensions (D x W x H) Interior 14.6 x 39.5 x 60.5 in.
Product Type Combi Safety Storage Cabinet
Dimensions (D x W x H) 18 x 43 x 65 in.
Load Bearing Capacity 45 gal. (170 L)
No. of Shelves 2
No. of Doors 2
Color Royal Blue