missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
GREENER_CHOICE

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

Supplier:  Justrite™ 896000

 View more versions of this product

Catalog No. 191202337


Only null left

Description

Description

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements

Certifications

FM approved except 19-120-2336.

Compliance

Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.

Warranty

10-Year

Specifications

Chemical Identifiers

Yellow
Can Storage Cabinet
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
Epoxy, Polyester
Steel (CR)
Flammable liquid storage and protection
2

Specifications

30.34 x 30.75 x 60.5 in.
165.1 cm
60 gal.
2
78 cm
2
34 in.
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
310 lb.
Manual-close
FM, NFPA, OSHA
60.5 in.
Steel (CR)
Can Storage Cabinet
153.7 cm
141 kg
77.1 cm
86.4 cm
86.4 cm
30.34 in.
65 in.
34 in.
Flammable liquid storage and protection
Yellow
Epoxy, Polyester
30.75 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions