missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Nonmetallic Dispensing Can

Catalog No. 17190B
Change view
Click to view available options
Capacity (Metric):
0.24 L
0.95 L
Color:
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color
17190B 0.95 L Red
17190A 0.24 L Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17190B Supplier Justrite™ Supplier No. 14010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.

  • Ideal for use with flammables
  • Leakproof, self-closing dispenser valves minimize spills, controls excess
  • High-density polyethylene body
  • Permits easy, one-hand dispensing of hazardous liquids directly onto work surface

Specifications

Certifications/Compliance FM
Product Type Dispensing Can
Capacity (English) 1 qt.
Capacity (Metric) 0.95 L
Color Red
For Use With (Application) For use with Flammables
Material Polyethylene
Includes Leakproof Brass Dispenser Valves Self-close
Diameter (English) Outer 4 in.
Diameter (Metric) Outer 10.1 mm