missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Nonmetallic Dispensing Can

Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.

Supplier:  Justrite™ 14004

 View more versions of this product

Catalog No. 17190A


Only null left

Description

Description

  • Ideal for use with flammables
  • Leakproof, self-closing dispenser valves minimize spills, controls excess
  • High-density polyethylene body
  • Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications

Chemical Identifiers

Dispensing Can
For use with Flammables
Polyethylene
Leakproof Brass Dispenser Valves Self-close

Specifications

FM
0.5 pt.
Red
Polyethylene
3.1 in.
Dispensing Can
0.24 L
For use with Flammables
Leakproof Brass Dispenser Valves Self-close
7.8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions