Learn More
Justrite™ Maintenance Spill Berms
To contain minor spills and leaks during temporary storage of drums, machine parts, batteries and equipment
$597.96 - $2,198.45
| Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
|
19111014
|
Justrite™
28400 |
10 gal. | 0.6 x 0.6 x 0.1 m |
Each for $1,259.48
|
|
||||
|
19111015
|
Justrite™
28402 |
20 gal. | 1.2 x 0.6 x 0.1 m |
Each for $1,232.50
|
|
||||
|
19111016
|
Justrite™
28404 |
40 gal. | 2.4 x 0.6 x 0.1 m |
Each for $1,474.31
|
|
||||
|
19111017
|
Justrite™
28406 |
40 gal. | 1.2 x 1.2 x 0.1 m |
Each for $1,515.59
|
|
||||
|
19111018
|
Justrite™
28408 |
60 gal. | 1.8 x 1.2 x 0.1 m |
Each for $1,571.18
|
|
||||
|
19111019
|
Justrite™
28410 |
80 gal. | 2.4 x 1.2 x 0.1 m |
Each for $1,750.73
|
|
||||
|
19111020
|
Justrite™
JMC28412 |
90 gal. | 1.8 x 1.8 x 0.1 m |
Each for $1,826.96
|
|
||||
|
19111021
|
Justrite™
28414 |
5 gal. | 0.6 x 0.6 x 0.05 m |
Each for $597.96
|
|
||||
|
19111022
|
Justrite™
JMC28416 |
10 gal. | 1.2 x 0.6 x 0.05 m |
Each for $719.45
|
|
||||
|
19111024
|
Justrite™
28421 |
40 gal. | 2.4 x 1.2 x 0.05 m |
Each for $1,000.15
|
|
||||
|
19111023
|
Justrite™
28418 |
20 gal. | 1.2 x 1.2 x 0.05 m |
Each for $890.40
|
|
||||
|
19111025
|
Justrite™
28422 |
44 gal. | 1.8 x 1.8 x 0.05 m |
Each for $1,236.85
|
|
||||
|
19111027
|
Justrite™
28426 |
220 gal. | 5.4 x 3 x 0.05 m |
Each for $2,198.45
|
|
||||
|
19111026
|
Justrite™
28424 |
135 gal. | 3.3 x 3.04 x 0.05 m |
Each for $1,390.06
|
|
||||
Description
- Made of broad UV and chemical resistant PVC coated fabric
- No assembly required
- Berm anchored in place using brass grommets in each corner
- Encased urethane foam sidewalls for easy roll-through access without need for ramp
- Walls spring back into place without memory loss
- Anchor stakes not included
- Optional grates to keep products and machinery above fluid can be ordered separately