missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Flammables Two-Door Wall-Mount Cabinet
GREENER_CHOICE

Catalog No. 1798560
Change view
Click to view available options
Load Bearing Capacity:
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity
1798560 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1798560 Supplier Justrite™ Supplier No. 893400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Five adjustable shelves and mounting hardware.

Safe and convenient storage of small containers of liquid flammables

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Can be wall mounted or placed on a workbench to keep floor space free

Certifications

FM approved.

Compliance

Meets OSHA and NFPA requirements.

Warranty

10 years

Specifications

Dimensions (D x W x H) Interior 8.75 x 39.5 x 39.62 in.
No. of Doors 2
No. of Shelves 5
Load Bearing Capacity 20 gal.
Product Type Wall mount storage cabinet
Height (Metric) Interior 100.6 cm
Depth (Metric) Exterior 30.5 cm
Height (Metric) Exterior 112 cm
Weight (Metric) 93 kg
Width (Metric) Exterior 109 cm
Depth (Metric) Interior 22.2 cm
Width (Metric) Interior 100.3 cm
Height (English) Exterior 44 in.
Depth (English) Exterior 12 in.
Weight (English) 205 lb.
Width (English) Interior 39.5 in.
Height (English) Interior 39.62 in.
Material Steel (CR)
Depth (English) Interior 8.75 in.
Width (English) Exterior 43 in.
Color Yellow
For Use With (Application) Flammables
Certifications/Compliance FM, NFPA, OSHA
Door Style Manual-close
Surface Coating Epoxy, Polyester
Includes Five adjustable shelves and mounting hardware, U-Loc™ handles with two keys, Haz-Alert™ reflective trilingual exterior labeling and Galvanized Steel SpillSlope™ shelves that direct all spills to the 5.
Show More Show Less