missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ EX Flammable Safety Cabinet

Description
Features of Justrite Flammables Cabinets
- Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Lettering on door reads: FLAMMABLE—KEEP FIRE AWAY
- Accepts padlock (not included)
- Color: yellow, gray (optional), white (19-160-376)
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and comply with OSHA regulations.
Warranty
10 years
Specifications
Specifications
| Dimensions (D x W x H) Interior | 14.56 x 39.5 x 60.5 in. |
| Product Type | Can Storage Cabinet |
| Load Bearing Capacity | 45 gal. |
| Certifications/Compliance | FM, NFPA, OSHA |
| Width (Metric) Exterior | 109 cm |
| Depth (Metric) Exterior | 457 cm |
| Height (Metric) Exterior | 165.1 cm |
| Depth (Metric) Interior | 37 cm |
| Weight (Metric) | 136 kg |
| Width (Metric) Interior | 100.3 cm |
| Show More |