missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ Piggyback Flammable Safety Cabinet
Designed to mount on top of 30- and 45 gallon Sure-Grip™ EX cabinets, these Piggyback safety cabinets can be used independently or stacked as benchtop units for added storage.
Supplier: Justrite™ 891300
Description
Piggyback Safety Cabinet Features
- Stack atop standard 30 gal. (114L) safety cabinets
- Can also be wall mounted or used on countertop
- 18-gauge steel with 1.5 in. (3.8cm) insulating air space
- Available with manual or self-closing doors with flush-mounted Sure-Grip™ handles with exclusive gripper pad for secure hold and security
Features of Justrite Flammables Cabinets
- Double-wall construction provides 1 1/2 in. of insulating air space to help prevent the spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Backed by an exclusive 10-year warranty
- Meet OSHA and NFPA requirements
Certifications
FM approved. Self-closing models also UL listed.
Compliance
Comply with Uniform Fire Code.
Chemical Identifiers
| 14.56 in. | |
| 37 cm | |
| 18 in. | |
| Piggyback Storage Cabinet | |
| Flammables | |
| 2 | |
| 39.5 in. | |
| 100.3 cm |
| 45.7 cm | |
| Yellow | |
| Steel (CR) | |
| 43 in. | |
| Flush mounted Sure-Grip™ handles with cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, dual vents with flame arresters, four adjustable self leveling feet, grounding con | |
| Epoxy, Polyester | |
| 109 cm |
Specifications
| 14.56 x 39.5 x 13 in. | |
| 12 gal. | |
| Flammables | |
| 43 in. | |
| 18 in. | |
| Epoxy, Polyester | |
| 45.7 cm | |
| 2 | |
| Flush mounted Sure-Grip™ handles with cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, dual vents with flame arresters, four adjustable self leveling feet, grounding con | |
| Steel (CR) | |
| Manual-close | |
| 129 lb. | |
| 14.56 in. |
| Piggyback Storage Cabinet | |
| 37 cm | |
| 59 kg | |
| 33 cm | |
| 100.3 cm | |
| 18 in. | |
| 109 cm | |
| 45.7 cm | |
| 13 in. | |
| Yellow | |
| FM, NFPA, OSHA | |
| 39.5 in. |