missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Compac Flammable Safety Cabinet
GREENER_CHOICE

Catalog No. p-27058
Change view
Click to view available options
Door Style:
Manual-close
Self-close
Weight (English):
120 lb.
131 lb.
Weight (Metric):
54.4 kg
69 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style Weight (English) Weight (Metric)
19121853 Self-close 131 lb. 69 kg
19121852 Manual-close 120 lb. 54.4 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19121853 Supplier Justrite™ Supplier No. 891220

Includes

One adjustable shelf.

Justrite Compac Safety Cabinets fit nearly anywhere and provide protection where space is at a premium. The cabinets protect workers, reduce fire risks, and allow storage of flammable fuels and chemicals in code-compliant safety cabinets.

  • One-door cabinet stores up to 12 gal. (45.4L) of flammables
  • Double-wall construction provides 11/2 in. of insulating air space to help prevent the spread of flames Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • High-visibility Haz-Alert™ reflective warning labels provide added safety during power outages or fires
  • Self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • One adjustable SpillSlope™ safety shelf directs spills to back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Finished inside and out with a durable lead-free hybrid epoxy and polyester powder paint for chemical resistance
  • Fit neatly under counter or stand alone
  • Lettering on door reads: FLAMMABLE—KEEP FIRE AWAY
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements

19-121-852: Manual-close door

19-121-853; Self-closing door

Certifications

FM approved. Self-closing model UL listed.

Compliance

Meet NFPA and OSHA requirements. Self-closing model complies with Uniform Fire Code.

Specifications

Dimensions (D x W x H) Interior 14.56 x 20 x 30.5 in.
Product Type Flammables Storage Cabinet
Load Bearing Capacity 12 gal.
Door Style Self-close
Depth (Metric) Exterior 45.7 cm
Weight (Metric) 69 kg
Width (Metric) Exterior 59.05 cm
Width (Metric) Interior 50.8 cm
No. of Doors 1
Height (Metric) Interior 77.5 cm
No. of Shelves 1
Depth (Metric) Interior 37 cm
Height (Metric) Exterior 88.9 cm
Includes One adjustable shelf, U-Loc™ handle with two keys and accept padlock, Haz-Alert™ reflective exterior labeling and Galvanized Steel SpillSlope™ shelves, leakproof 2 in. bottom sump, dual vents with flam
Width (English) Exterior 23.25 in.
Color Yellow
For Use With (Application) Flammables
Width (English) Interior 20 in.
Height (English) Exterior 35 in.
Height (English) Interior 30.5 in.
Weight (English) 131 lb.
Depth (English) Exterior 18 in.
Certifications/Compliance FM, NFPA, OSHA
Depth (English) Interior 14.56 in.
Material Steel (CR)
Surface Coating Epoxy, Polyester
Show More Show Less