Learn More
Justrite™ Extra Shelves for Locker, Accessory
Used as additional shelf with existing shelving system or replace damaged one
Supplier: Justrite™ 915122
Description
- Movable
- Does not come with wall mount unistrut
- Not for use in locker models with explosion-relief panels
Chemical Identifiers
| Galvanized Steel | |
| Safety locker | |
| 40.6 cm |
| Extra Shelf | |
| 16 in. |
Specifications
| Galvanized Steel | |
| 1.2 m | |
| 4 ft. | |
| 16 in. |
| Extra Shelf | |
| 40.6 cm | |
| Safety locker |