Learn More
Justrite™ Drum Faucets, Extender, and Valve
Self-closing safety drum faucets
Supplier:  Justrite™ 08902
 View more versions of this product
Catalog No. 1798654
Only null left
Description
Description
  • Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
  • Brass faucets screw into drum; adjustable model adjusts to spout-down position
  • Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
  • Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Chemical Identifiers
FM
Specifications
FM
Rigid Drum Faucet
Brass
Teflon Seal, Spout Downward
Product Suggestions
Product Suggestions