Learn More
Justrite™ Drum Faucets, Extender, and Valve
Catalog No. p-33603
Change view
Click to view available options
Product Type:
Adjustable Drum Faucet
Drum Gate Valve
Flexible Hose Extension
Rigid Drum Faucet
Material:
Brass
Stainless Steel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
19812498 Flexible Hose Extension Stainless Steel
17185C Flexible Hose Extension Brass
17185D Drum Gate Valve Brass
1798653 Adjustable Drum Faucet Brass
1798654 Rigid Drum Faucet Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19812498 Supplier Justrite™ Supplier No. 08587
Only null left
Self-closing safety drum faucets
  • Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
  • Brass faucets screw into drum; adjustable model adjusts to spout-down position
  • Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
  • Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Certifications/Compliance FM
Product Type Flexible Hose Extension
For Use With (Application) Provide Extra Accuracy and Prevent Spills
For Use With (Equipment) 08916 Faucet
Material Stainless Steel
Height (English) 6 in.
Height (Metric) 15.2 cm