missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Dispensing Safety Cans
GREENER_CHOICE

Catalog No. 19413849
Change view
Click to view available options
Product Type:
Type I Shelf Safety Can
Type I Tilt Safety Can
Height (English):
15.8 in.
16.8 in.
Height (Metric):
403 mm
429 mm
Includes:
Tap 08540
Tap 08902
Tap 08902 and Stand
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Height (English) Height (Metric) Includes
19413849 Type I Tilt Safety Can 15.8 in. 403 mm Tap 08902 and Stand
19413847 Type I Shelf Safety Can 16.8 in. 429 mm Tap 08540
19413848 Type I Shelf Safety Can 16.8 in. 429 mm Tap 08902
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19413849 Supplier Justrite™ Supplier No. 7150156
Only null left

Reliable dispensing features at an economical price

For safely dispensing hazardous liquids into test tubes, small flasks and beakers

  • Vacuum-relief valve within the lid for smooth liquid flow and self-closing faucets for trouble-free dispensing
  • Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
  • Capacity: 19L (5 gal)
  • Color: Red
  • Smaller 19-413-847 faucet offers easier dispensing in lab environments; No. 08902 faucet better suited for heavy, industrial use

Shelf Cans

  • Faucet mounted two inches above the bottom to eliminate lifting the cans during dispensing

Tilt Cans

  • Faucet on top with a sturdy metal cradle for the can; cradle base measures 35.1 x 35.6cm (13.81 x 14 in.)

Certifications

FM

Compliance

CARB, NFPA, OSHA

Specifications

Certifications/Compliance FM, CARB, NFPA, OSHA
Product Type Type I Tilt Safety Can
Closure Type Self Closing Pressure Relief Cap
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Color Red
For Use With (Application) For Laboratories
Material Steel
Includes Tap 08902 and Stand
Height (English) 15.8 in.
Height (Metric) 403 mm
Diameter (English) Inner 11.75 in.
Diameter (English) Opening 1.58 in.
Diameter (Metric) Inner 298 mm
Diameter (Metric) Opening 4 cm
Show More Show Less