missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Chemcor™ Lined Safety Cabinets for Hazardous Materials

Give your workers the edge. Justrite ChemCor™ thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals.
Specifications
| Color | Blue |
|---|---|
| Product Type | Lined Safety Cabinet |
| Door Style | Self-close |
| For Use With (Application) | Hazardous materials |
| No. of Doors | 2 |
| Catalog Number | Mfr. No. | Load Bearing Capacity | No. of Shelves | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Load Bearing Capacity | No. of Shelves | Price | Quantity & Availability | ||||
19414811
![]() |
Justrite™
8645282 |
170 L | 2 |
N/A
|
|
||||
19414810
![]() |
Justrite™
8630282 |
113.5 L | 1 |
N/A
|
|
||||
19414812
![]() |
Justrite™
8660282 |
227 L | 2 |
N/A
|
|
||||
Description
- Protection against corrosion from drips and off-gassing from harsh chemicals
- Feature unique, flame-coated thermoplastic finish on interior doors, shelves, walls, ceiling, and sump
- Non-porous, seam-free surface eliminates need for corrosion-prone rivets or fasteners
- Resists staining and cleans up easily
- Includes separate polyethylene work tray that can be placed on top
Specifications
| Blue | |
| Self-close | |
| 2 |
| Lined Safety Cabinet | |
| Hazardous materials |