Learn More
Justrite™ Chemcor™ Lined Safety Cabinets for Hazardous Materials
GREENER_CHOICE
Give your workers the edge. Justrite ChemCor™ thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals.
Supplier:  Justrite™ 8630282
 View more versions of this product
Catalog No. 19414810
Only null left
Description
Description
  • Protection against corrosion from drips and off-gassing from harsh chemicals
  • Feature unique, flame-coated thermoplastic finish on interior doors, shelves, walls, ceiling, and sump
  • Non-porous, seam-free surface eliminates need for corrosion-prone rivets or fasteners
  • Resists staining and cleans up easily
  • Includes separate polyethylene work tray that can be placed on top
Specifications
Chemical Identifiers
Blue
Self-close
2
Lined Safety Cabinet
Hazardous materials
Specifications
Self-close
113.5 L
1
Blue
Lined Safety Cabinet
2
Hazardous materials
Product Suggestions
Product Suggestions