missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Antistatic Wires with End Connectors

Prevent ignition by sparks resulting from movement of flammable liquids

$124.10 - $147.97

Specifications

Product Type Wire
View More Specs

Products 4
Catalog Number Mfr. No. Includes Material Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Includes Material Length (Metric) Price Quantity & Availability  
1798590A
View Documents Promotion Details
Justrite™
08496
End Connectors Zinc, Steel 304.8 cm
Each for $136.64
Only null left
 
1798590B
View Documents Promotion Details
Justrite™
08497
End Connectors Copper, Polyurethane 304.8 cm
Each for $124.10
Only null left
 
1798590F
View Documents Promotion Details
Justrite™
08501
C Clamp, Alligator Clip Zinc, Steel 91.44 cm
Each for $124.34
Only null left
 
1798590H
View Documents Promotion Details
Justrite™
08505
Insulated Wire, C Clamp, Alligator Clip Zinc, Steel 91.44 cm
Each for $147.97
Only null left
 
Description

Description

Choose from flexible or insulated wire.

  • Flexible wire constructed of multistrand aircraft cable encased in zinc-coated steel
  • Insulated wire constructed of multistrand copper wire with black polyurethane electricity-resistant coating
  • Standard grounding wire lengths are 3 and 10 ft. (1 and 3m); when longer lengths are needed, use several wires with 0.25 in. terminal ends (Cat. Nos. 17-985-90G or 17-985-90J) bolted together

†Coiled.

Compliance

Comply with OSHA standards and local safety codes requiring grounded containers during the transfer of liquids and drums connected to an earth ground.

Specifications

Specifications

Wire
Videos
SDS
Documents

Documents