missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Jupiter Electronic Pipette, 8-Channel

Catalog No. S00058
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
15 to 300 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S00058 15 to 300 μL
S00056 0.5 to 10 μL
S00057 10 to 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Prop 65
Catalog No. S00058 Supplier United Scientific™ Supplier No. UNUNEMCH0300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prop 65

Versatile multi-channel pipettes deliver fast and accurate results with three operating modes: pipetting, step dispensing, and diluting.

  • Increase efficiency and speed with the Jupiter 8-Channel Electronic Pipettes designed for precise, flexible liquid handling.
  • Supports multiple functions including pipetting, repetitive dispensing, and diluting.
  • Simple two-knob interface and clear digital display allow quick switching between modes, volumes, and speeds.
  • High-performance stepper motor ensures accuracy, repeatability, and adjustable aspiration/dispense speeds.
  • Lightweight and well-balanced pipette reduces fatigue during extended use; compatible with most universal tip brands.
  • Intuitive LCD interface and 360° rotating tip ejector collar for smooth, comfortable handling.
  • Long-lasting lithium-ion battery recharges via AC adapter or USB; includes wall-mounted charging stand.
  • Calibrated and tested in compliance with ISO8655-6 / DIN12650 standards.
  • Available in multiple volume capacities.

Specifications

Battery Type Lithium Ion
Certifications/Compliance ISO8655-6 / DIN12650
Compatible Tips 300, 350 μL
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette Tip 300, 350 μL
Grip Type Ergonomic
Material Plastic
Model Jupiter
Number of Channels 8
Programmability No
Sterility Non-sterile
Type Multichannel Pipette with Variable Volume Setting
Volume (Metric) 15 to 300 μL
Display Digital
Increments 1.0 μL
Show More Show Less