missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Jump'n Jacks DFS Item

$40.77 - $62.27

Specifications

Product Type Canine Toy
Color Red
Material Rubber
For Use With (Application) Cleaning and conditioning of teeth and gums
View More Specs

Products 2
Catalog Number Mfr. No. Size Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Size Dimensions (L x W) Price Quantity  
14727016 DFS Item
View Documents
Bio-Serv™
K3540
Small 2.75 x 2.75 in. N/A
Only null left
 
14727017 DFS Item
View Documents
Bio-Serv™
K3541
Medium 3.5 x 3.5 in. N/A
Only null left
 
Description

Description

  • Made of flexible natural rubber
  • Unique shape minimizes loss through cage
  • Ribbed texture promotes natural play activity
  • Promotes good dental health
  • Cagewasher safe
  • Red
Specifications

Specifications

Canine Toy
Red
Rubber
Cleaning and conditioning of teeth and gums
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'