missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Jump'n Jacks

Catalog No. 14727016
Encompass
Change view
Click to view available options
Size:
Small
Medium
Dimensions (L x W):
2.75 x 2.75 in.
3.5 x 3.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Dimensions (L x W)
14727016 Small 2.75 x 2.75 in.
14727017 Medium 3.5 x 3.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14727016 Supplier Bio-Serv™ Supplier No. K3540
  • Made of flexible natural rubber
  • Unique shape minimizes loss through cage
  • Ribbed texture promotes natural play activity
  • Promotes good dental health
  • Cagewasher safe
  • Red

Specifications

Product Type Canine Toy
Color Red
Material Rubber
For Use With (Application) Cleaning and conditioning of teeth and gums
Size Small
Dimensions (L x W) 2.75 x 2.75 in.