missing translation for 'onlineSavingsMsg'
Learn More

Julolidine 97.0+%, TCI America™

Catalog No. p-7131734
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J00075G 5 g
J000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. J00075G Supplier TCI America Supplier No. J00075G
Only null left

Chemical Identifiers

CAS 479-59-4
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
InChI Key DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Synonym julolidine, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine, 1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, 2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline, 2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
PubChem CID 68069
SMILES C1CC2=C3C(=CC=C2)CCCN3C1

Specifications

Melting Point 37°C
Boiling Point 280°C
Quantity 5 g
Formula Weight 173.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Julolidine
TSCA Yes
Recommended Storage Refrigerator