missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520640
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520640 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520640 Supplier Invitrogen™ Supplier No. PA520640
Only null left

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20640 can be used with blocking peptide PEP-0760.

Junctional complexes between the plasma membrane and endoplasmic/sarcoplasmic reticulum are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. The protein encoded by this gene is a component of junctional complexes and is composed of a C-terminal hydrophobic segment spanning the endoplasmic/sarcoplasmic reticulum membrane and a remaining cytoplasmic domain that shows specific affinity for the plasma membrane. This gene is a member of the junctophilin gene family.

Specifications

Antigen JPH1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Jph1
Gene Accession No. Q9ET80, Q9HDC5
Gene Alias CMT2K; ENSMUSG00000054314; JP1; JP-1; Jph1; junctophilin 1; junctophilin type 1; junctophilin type1; Junctophilin-1; mitsugumin72
Gene Symbols Jph1
Host Species Rabbit
Immunogen A 15 amino acid peptide near the carboxy terminus of human JPH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297748, 56704, 57339
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less