missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jouberin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570269 Supplier Invitrogen™ Supplier No. PA570269

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen Jouberin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AHI1
Gene Accession No. Q6DTM3
Gene Alias 1700015F03Rik; Abelson helper integration site 1; abelson helper integration site 1 protein; abelson helper integration site 1 protein homolog; Ahi1; AHI-1; contatins SH3 and WD40 domains; D10Bwg0629e; dJ71N10.1; JBTS3; jouberin; ORF1; RP1-32B1.2
Gene Symbols AHI1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat Jouberin (aa 671-720).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308923
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less