missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JMJD2B Recombinant Superclonal™ Antibody (9HCLC)

Catalog No. 712584
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712584 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 712584 Supplier Invitrogen™ Supplier No. 712584
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt].

Specifications

Antigen JMJD2B
Applications Western Blot
Classification Recombinant Superclonal
Clone 9HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene KDM4B
Gene Accession No. O94953
Gene Alias 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B
Gene Symbols KDM4B
Host Species Rabbit
Immunogen Protein corresponding to human JMJD2B [aa1031-aa1096].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less