missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JLP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | JLP |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Extracellular signals are transduced into cells through mitogen-activated protein kinases. The structural organization of these kinases into specific signaling domains is facilitated by scaffolding proteins involved in closely tethering different kinases so that successive phosphorylation events can occur. The protein encoded by this gene is a scaffolding protein that brings together mitogen-activated protein kinases and their transcription factor targets for the activation of specific signaling pathways. This gene which is abundantly expressed in testicular haploid germ cells encodes a protein that is recognized by sperm-agglutinating antibodies and implicated in infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| JLP | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| C-jun-amino-terminal kinase-interacting protein 4; JNK-interacting protein 4; JIP-4; JNK-associated leucine-zipper protein; JLP; Sperm-associated antigen 9; Mitogen-activated protein kinase 8-interacting protein 4; Human lung cancer protein 6; HLC-6; Proliferation-inducing protein 6; Sperm-specific protein; Sperm surface protein; Protein highly expressed in testis; PHET; Sunday driver 1; Cancer/testis antigen 89; CT89; SPAG9; HSS; KIAA0516; MAPK8IP4; SYD1 | |
| Spag9 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60271, Q58A65 | |
| 360600, 70834, 9043 | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| SPAG9 |