Learn More
Fisherbrand™ Surface Treated SterileTissue Culture Plates
Ideal for comparison and analysis of cell growth, yield in multiple platforms.
Catalog No. FB012929
Only null left
Explore available promotions
Description
Description
  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Warranty
Three years
Specifications
Specifications
4 x 6
Flat
Natural
Polystyrene
Sterile
Cell Culture Plate
Tissue culture
Individually wrapped
CE
3.5 mL
1.93 cm2
Rectangular
Standard
24
100 Plates
Product Suggestions
Product Suggestions