missing translation for 'onlineSavingsMsg'
Learn More

Jervine 97.0+%, TCI America™

Catalog No. J000910MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J000910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J000910MG Supplier TCI America Supplier No. J000910MG
Only null left

Chemical Identifiers

CAS 469-59-0
Molecular Formula C27H39NO3
Molecular Weight (g/mol) 425.61
MDL Number MFCD01684066
InChI Key CLEXYFLHGFJONT-UHFFFAOYNA-N
Synonym jervine, iervin, jerwiny polish, 11-ketocyclopamine, unii-19v3ecx465, veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3beta,23beta, 17,23beta-epoxy-3beta-hydroxyveratraman-11-one, spiro 9h-benzo a fluorene-9,2' 3'h-furo 3,2-b-pyridin-11 1h-one, 2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11a,11b-hexadecahydro-3-hydroxy-3',6',10, 11b-tetramethyl, jerwiny, veratraman-11-one, 17,23-epoxy-3-hydroxy-, 3-beta,23-beta
PubChem CID 10098
ChEBI CHEBI:6088
IUPAC Name 3-hydroxy-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-11-one
SMILES CC1C2NCC(C)CC2OC11CCC2C3CC=C4CC(O)CCC4(C)C3C(=O)C2=C1C

Specifications

Melting Point 246°C
Color White-Yellow
Quantity 10 mg
UN Number 2811
Formula Weight 425.61
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Jervine
RTECSNumber WG9700000
TSCA No
Recommended Storage Freezer