accessibility menu, dialog, popup

UserName

CAS RN 469-59-0

H3CHNCH3OHH3COH3CO

CAS RN 469-59-0

IUPAC Name: 3-hydroxy-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-11-one
Synonyms: jervine
Molecular Weight (g/mol): 425.61
Molecular Formula: C27H39NO3
InChi Key: CLEXYFLHGFJONT-UHFFFAOYNA-N
SMILES: CC1C2NCC(C)CC2OC11CCC2C3CC=C4CC(O)CCC4(C)C3C(=O)C2=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results