missing translation for 'onlineSavingsMsg'
Learn More

Jasmonic Acid (mixture of isomers) 85.0+%, TCI America™

Catalog No. J00045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J00045G 5 g
J00041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. J00045G Supplier TCI America Supplier No. J00045G

Chemical Identifiers

CAS 221682-41-3
Molecular Formula C12H18O3
Molecular Weight (g/mol) 210.273
InChI Key ZNJFBWYDHIGLCU-ARJAWSKDSA-N
Synonym 3-(Carboxymethyl)-2-(2-pentenyl)cyclopentanone, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid
PubChem CID 12304694
IUPAC Name 2-[3-oxo-2-[(Z)-pent-2-enyl]cyclopentyl]acetic acid
SMILES CCC=CCC1C(CCC1=O)CC(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 210.27
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Jasmonic Acid (mixture of isomers)